2,6-Difluoro-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2,6-Difluoro-4-methoxy-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502825-1G
APOSPC502825-1GEA 1180 EUR
APOSPC502825-1G
2,6-Difluoro-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2,6-Difluoro-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₇F₇O₂
Molecular Weight: 340,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630099
CAS-Nummer: 2244084-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR