3-Bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Bromophenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502827-1G
APOSPC502827-1GEA 1180 EUR
APOSPC502827-1G
3-Bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆BrF₅O
Molecular Weight: 353,082 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629953
CAS-Nummer: 2244085-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR