4-Bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
PC502828-1G
APOSPC502828-1GEA
1180
EUR
APOSPC502828-1G
4-Bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₆BrF₅O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629957 CAS-Nummer: 2244086-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...