2-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Chlorophenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502829-1G
APOSPC502829-1GEA 1180 EUR
APOSPC502829-1G
2-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆ClF₅O
Molecular Weight: 308,631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629961
CAS-Nummer: 2244086-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR