3-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502830-1G
APOSPC502830-1GEA 1180 EUR
APOSPC502830-1G
3-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆ClF₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD31629965
CAS-Nummer: 2244084-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR