2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluorophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(4-fluorophenoxy)-5-(trifluoromethyl)benzene

PC502832-1G
APOSPC502832-1GEA 1180 EUR
APOSPC502832-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluorophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluorophenyl ether
Formel: C₁₃H₆F₆O
Molecular Weight: 292,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629971
CAS-Nummer: 2244084-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR