2,6-Difluoro-4-(trifluoromethyl)phenyl 2-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(2-nitrophenoxy)-5-(trifluoromethyl)benzene

PC502833-1G
APOSPC502833-1GEA 1180 EUR
APOSPC502833-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-nitrophenyl ether
Formel: C₁₃H₆F₅NO₃
Molecular Weight: 319,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629974
CAS-Nummer: 2244083-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR