2,6-Difluoro-4-(trifluoromethyl)phenyl 4-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(4-nitrophenoxy)-5-(trifluoromethyl)benzene

PC502835-1G
APOSPC502835-1GEA 1180 EUR
APOSPC502835-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-nitrophenyl ether
Formel: C₁₃H₆F₅NO₃
Molecular Weight: 319,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629980
CAS-Nummer: 2244088-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR