2,6-Difluoro-4-(trifluoromethyl)phenyl 3-methoxyphenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(3-methoxyphenoxy)-5-(trifluoromethyl)benzene

PC502836-1G
APOSPC502836-1GEA 1180 EUR
APOSPC502836-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-methoxyphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-methoxyphenyl ether
Formel: C₁₄H₉F₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31629983
CAS-Nummer: 2244087-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR