2-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502838-1G
APOSPC502838-1GEA 1180 EUR
APOSPC502838-1G
2-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₈F₅NO
Molecular Weight: 289,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629989
CAS-Nummer: 2244085-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR