4-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502840-1G
APOSPC502840-1GEA 1180 EUR
APOSPC502840-1G
4-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₈F₅NO
Molecular Weight: 289,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629995
CAS-Nummer: 2244083-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR