2-Amino-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-5-fluoroaniline
PC502841-1G
APOSPC502841-1GEA
1760
EUR
APOSPC502841-1G
2-Amino-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₇F₆NO Molecular Weight: 307,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629998 CAS-Nummer: 2244088-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...