5-Amino-2-chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline

PC502843-1G
APOSPC502843-1GEA 1760 EUR
APOSPC502843-1G
5-Amino-2-chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2-chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆ClF₆NO
Molecular Weight: 341,636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630004
CAS-Nummer: 2244086-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR