2-Amino-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4-fluoroaniline

PC502844-1G
APOSPC502844-1GEA 1760 EUR
APOSPC502844-1G
2-Amino-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇F₆NO
Molecular Weight: 307,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630007
CAS-Nummer: 2244088-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR