2-Amino-6-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502845-1G
APOSPC502845-1GEA 1760 EUR
APOSPC502845-1G
2-Amino-6-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-6-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇BrF₅NO
Molecular Weight: 368,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630010
CAS-Nummer: 2244085-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR