2-Amino-4-bromo-6-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-3-chloro-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502846-1G
APOSPC502846-1GEA 1760 EUR
APOSPC502846-1G
2-Amino-4-bromo-6-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-bromo-6-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆BrClF₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31630013
CAS-Nummer: 2244088-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR