2-Amino-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-6-fluoroaniline

PC502847-1G
APOSPC502847-1GEA 1760 EUR
APOSPC502847-1G
2-Amino-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇F₆NO
Molecular Weight: 307,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630016
CAS-Nummer: 2244086-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR