2-Amino-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-5-methoxyaniline
PC502848-1G
APOSPC502848-1GEA
1760
EUR
APOSPC502848-1G
2-Amino-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₄H₁₀F₅NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630019 CAS-Nummer: 2244085-46-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...