5-Amino-2-bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline

PC502849-1G
APOSPC502849-1GEA 1760 EUR
APOSPC502849-1G
5-Amino-2-bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2-bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆BrF₆NO
Molecular Weight: 386,087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630022
CAS-Nummer: 2244088-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR