2-Amino-4-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502850-1G
APOSPC502850-1GEA 1760 EUR
APOSPC502850-1G
2-Amino-4-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇BrF₅NO
Molecular Weight: 368,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630025
CAS-Nummer: 2244086-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR