2-Amino-6-bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-5-fluoroaniline
PC502853-1G
APOSPC502853-1GEA
1760
EUR
APOSPC502853-1G
2-Amino-6-bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-6-bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₆BrF₆NO Molecular Weight: 386,087 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630034 CAS-Nummer: 2244083-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...