3-Amino-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 3-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-2-methylaniline

PC502854-1G
APOSPC502854-1GEA 1760 EUR
APOSPC502854-1G
3-Amino-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Amino-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₁₀F₅NO
Molecular Weight: 303,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630037
CAS-Nummer: 2244087-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR