5-Amino-2-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-3-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502856-1G
APOSPC502856-1GEA 1760 EUR
APOSPC502856-1G
5-Amino-2-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇BrF₅NO
Molecular Weight: 368,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630043
CAS-Nummer: 2244085-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR