3-Amino-4-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline
PC502857-1G
APOSPC502857-1GEA
1760
EUR
APOSPC502857-1G
3-Amino-4-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Amino-4-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₇BrF₅NO Molecular Weight: 368,097 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630046 CAS-Nummer: 2244088-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...