2-Amino-3-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-6-methylaniline

PC502859-1G
APOSPC502859-1GEA 1760 EUR
APOSPC502859-1G
2-Amino-3-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-3-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₁₀F₅NO
Molecular Weight: 303,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630052
CAS-Nummer: 2244086-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR