5-Amino-2,4-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502860-1G
APOSPC502860-1GEA 1760 EUR
APOSPC502860-1G
5-Amino-2,4-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2,4-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆F₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31630055
CAS-Nummer: 2244086-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR