2-Amino-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-3-fluoroaniline

PC502861-1G
APOSPC502861-1GEA 1760 EUR
APOSPC502861-1G
2-Amino-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇F₆NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31630058
CAS-Nummer: 2244085-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR