2-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502862-1G
APOSPC502862-1GEA 1760 EUR
APOSPC502862-1G
2-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇BrF₅NO
Molecular Weight: 368,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630061
CAS-Nummer: 2244087-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR