2-Amino-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-5-methylaniline
PC502863-1G
APOSPC502863-1GEA
1760
EUR
APOSPC502863-1G
2-Amino-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₄H₁₀F₅NO Molecular Weight: 303,227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630064 CAS-Nummer: 2244087-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...