2-Amino-4-fluoro-5-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-5-fluoro-4-methylaniline

PC502864-1G
APOSPC502864-1GEA 1760 EUR
APOSPC502864-1G
2-Amino-4-fluoro-5-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-fluoro-5-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₉F₆NO
Molecular Weight: 321,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630067
CAS-Nummer: 2244087-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR