2-Amino-4,5-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4,5-Difluoroaniline

PC502865-1G
APOSPC502865-1GEA 1760 EUR
APOSPC502865-1G
2-Amino-4,5-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4,5-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆F₇NO
Molecular Weight: 325.182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630070
CAS-Nummer: 2244085-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR