6-Amino-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-3,4-Difluoroaniline

PC502866-1G
APOSPC502866-1GEA 1760 EUR
APOSPC502866-1G
6-Amino-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
6-Amino-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆F₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31630073
CAS-Nummer: 2244085-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR