2-Amino-5-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-6-fluoroaniline

PC502867-1G
APOSPC502867-1GEA 1760 EUR
APOSPC502867-1G
2-Amino-5-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-5-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆BrF₆NO
Molecular Weight: 386,087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630076
CAS-Nummer: 2244086-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR