5-Amino-2-fluoro-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502868-1G
APOSPC502868-1GEA 1760 EUR
APOSPC502868-1G
5-Amino-2-fluoro-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2-fluoro-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₉F₆NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31630079
CAS-Nummer: 2244085-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR