2-Amino-4-bromo-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-4-fluoroaniline

PC502869-1G
APOSPC502869-1GEA 1760 EUR
APOSPC502869-1G
2-Amino-4-bromo-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-bromo-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆BrF₆NO
Molecular Weight: 386.087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630082
CAS-Nummer: 2244088-78-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR