6-Amino-2-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-4-fluoroaniline
PC502870-1G
APOSPC502870-1GEA
1760
EUR
APOSPC502870-1G
6-Amino-2-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
6-Amino-2-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₆BrF₆NO Molecular Weight: 386,087 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630085 CAS-Nummer: 2244083-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...