2-Amino-4-bromo-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-3-fluoroaniline

PC502871-1G
APOSPC502871-1GEA 1760 EUR
APOSPC502871-1G
2-Amino-4-bromo-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4-bromo-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆BrF₆NO
Molecular Weight: 386.087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630088
CAS-Nummer: 2244087-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR