3-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502873-1G
APOSPC502873-1GEA 1760 EUR
APOSPC502873-1G
3-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₇BrF₅NO
Molecular Weight: 368,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630094
CAS-Nummer: 2244085-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR