5-Amino-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 3-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4-methoxyaniline

PC502874-1G
APOSPC502874-1GEA 1760 EUR
APOSPC502874-1G
5-Amino-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₁₀F₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31630097
CAS-Nummer: 2244084-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR