4-Amino-2,6-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-3,5-Difluoroaniline

PC502875-1G
APOSPC502875-1GEA 1760 EUR
APOSPC502875-1G
4-Amino-2,6-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Amino-2,6-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆F₇NO
Molecular Weight: 325,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630100
CAS-Nummer: 2244088-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR