5-Amino-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 5-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-2-fluoro-4-methylaniline

PC502876-1G
APOSPC502876-1GEA 1760 EUR
APOSPC502876-1G
5-Amino-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₉F₆NO
Molecular Weight: 321,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630103
CAS-Nummer: 2244083-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR