2-Amino-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4-iodoaniline
PC502877-1G
APOSPC502877-1GEA
1760
EUR
APOSPC502877-1G
2-Amino-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₇F₅INO Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629954 CAS-Nummer: 2244083-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...