2-Amino-4,5-dichlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4,5-Dichloro-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]aniline

PC502878-1G
APOSPC502878-1GEA 1760 EUR
APOSPC502878-1G
2-Amino-4,5-dichlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-4,5-dichlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₆Cl₂F₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31629958
CAS-Nummer: 2244085-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR