2-Amino-6-bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-5-(trifluoromethoxy)aniline

PC502879-1G
APOSPC502879-1GEA 1760 EUR
APOSPC502879-1G
2-Amino-6-bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-6-bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₆BrF₈NO₂
Molecular Weight: 452,094 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629962
CAS-Nummer: 2244086-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR