3-Bromo-5-methylbenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC502884-1G PC502884-250MG
APOSPC502884-1GEA 609 EUR
APOSPC502884-1G APOSPC502884-250MG
3-Bromo-5-methylbenzyl fluoride 95%
3-Bromo-5-methylbenzyl fluoride
Formel: C₈H₈BrF
Lagertemperatur: Raumtemperatur
MDL: MFCD31634829
CAS-Nummer: 2244087-79-2
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR