2-Bromo-3-nitrobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC502886-250MG
APOSPC502886-250MGEA 79.9 EUR
APOSPC502886-250MG
2-Bromo-3-nitrobenzyl fluoride 95%
2-Bromo-3-nitrobenzyl fluoride
Formel: C₇H₅BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31634831
CAS-Nummer: 2244083-36-9
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR