3-Bromo-5-fluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC502900-1G
APOSPC502900-1GEA 609 EUR
APOSPC502900-1G
3-Bromo-5-fluorobenzyl fluoride 95%
3-Bromo-5-fluorobenzyl fluoride
Formel: C₇H₅BrF₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30829524
CAS-Nummer: 1715032-20-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR