Mangan(II)fluorid 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Mangandifluorid
PC5030-5G
APOSPC5030-5GEA
0
EUR
APOSPC5030-5G
Mangan(II)fluorid 99%
Mangan(II)fluorid
Formel:
F₂Mn Schmelzpunkt: 856 °C Dichte: 3,98 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016222 CAS-Nummer: 7782-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...