4-Fluor-3-(isopropylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC5031-1G
APOSPC5031-1GEA 29 EUR
APOSPC5031-1G
4-Fluor-3-(isopropylcarbamoyl)phenylboronsäure 98%
4-Fluor-3-(isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Schmelzpunkt: 263…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08235040
CAS-Nummer: 874219-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR