3-(Cyclohexylcarbamoyl)-4-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (3-(Cyclohexylcarbamoyl)-4-fluorophenyl)boronic acid

PC5032-1G
APOSPC5032-1GEA 277 EUR
APOSPC5032-1G
3-(Cyclohexylcarbamoyl)-4-fluorphenylboronsäure 98%
3-(Cyclohexylcarbamoyl)-4-fluorphenylboronsäure
Formel: C₁₃H₁₇BFNO₃
Molecular Weight: 265,0900 g/mol
Schmelzpunkt: 231…233 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235043
CAS-Nummer: 874219-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR